Invitrogen
Manufacturer Code:PA542860
Catalog # PIPA542860
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ANP32E |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acidic (leucine-rich) nuclear phosphoprotein 32 family member E LANP-L LANPL MGC5350; acidic (leucine-rich) nuclear phosphoprotein 32 family, member E; Acidic leucine-rich nuclear phosphoprotein 32 family member E; Acidic leucine-rich nuclear phosphoprotein 32 family member E-like protein; Cerebellar postnatal development protein 1; LANP-L; LANP-like protein; leucine-rich acidic nuclear protein like
Gene Aliases: 2810018A15Rik; AI047746; AI326868; ANP32E; CPD1; LANP-L; LANPL; mLANP-L; wu:fb22e03; wu:fc04c12; wu:fc19a08; wu:fc79f04; wu:fi39c10; zgc:65995; zgc:85827
UniProt ID: (Human) Q9BTT0, (Mouse) P97822, (Rat) Q5XIE0, (Zebrafish) Q6NUW5
Entrez Gene ID: (Human) 81611, (Rabbit) 100345001, (Bovine) 507203, (Horse) 100054493, (Dog) 100855771, (Guinea pig) 100720019, (Mouse) 66471, (Rat) 361999, (Zebrafish) 406562
Molecular Function: enzyme modulator phosphatase inhibitor phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.