Invitrogen
Manufacturer Code:PA130729
Catalog # PIPA130729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (Human). (NP&lowbar112182) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acidic (leucine-rich) nuclear phosphoprotein 32 family, member E; Acidic leucine-rich nuclear phosphoprotein 32 family member E; Cerebellar postnatal development protein 1; LANP-L; LANP-like protein; leucine-rich acidic nuclear protein like; RP4-790G17.1 LANP-L LANPL MGC5350
Gene Aliases: 2810018A15Rik; AI047746; AI326868; ANP32E; CPD1; LANP-L; LANPL; mLANP-L
UniProt ID: (Human) Q9BTT0, (Mouse) P97822, (Rat) Q5XIE0
Entrez Gene ID: (Human) 81611, (Mouse) 66471, (Rat) 361999
Molecular Function:
enzyme modulator
phosphatase inhibitor
phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.