Invitrogen
Manufacturer Code:PA520026
Catalog # PIPA520026
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at carboxy terminus of human PHAP I. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid nuclear phosphoprotein 32 (leucine rich); acidic (leucine-rich) nuclear phosphoprotein 32 family, member A; acidic (leucine-rich) nuclear phosphoprotein 32 family, member E; Acidic leucine-rich nuclear phosphoprotein 32 family member A; Acidic leucine-rich nuclear phosphoprotein 32 family member E; acidic nuclear phosphoprotein 32; acidic nuclear phosphoprotein 32 family, member A; Acidic nuclear phosphoprotein pp32; C15orf1 I1PP2A LANP MAPM PHAP1 PHAPI PP32 acidic leucine-rich nuclear phosphoprotein 32 family member A acidic nuclear phosphoprotein pp32 cerebellar leucine rich acidic nuclear protein inhibitor-1 of protein phosphatase-2A leucine-rich acidic nuclear protein mapmodulin potent heat-stable protein phosphatase 2A inhibitor I1PP2A putative HLA-DR-associated protein I putative human HLA class II associated protein I; cerebellar leucine rich acidic nuclear protein; Cerebellar postnatal development protein 1; hepatopoietin Cn; inhibitor-1 of protein phosphatase-2A; LANP; LANP-L; LANP-like protein; Leucine-rich acidic nuclear protein; leucine-rich acidic nuclear protein LANP; leucine-rich acidic nuclear protein like; Mapmodulin; PHAPI; Potent heat-stable protein phosphatase 2A inhibitor I1PP2A; pp32; Putative HLA-DR-associated protein I; putative human HLA class II associated protein I; vacidic (leucine-rich) nuclear phosphoprotein 32 family, member A
Gene Aliases: 2810018A15Rik; AI047746; AI326868; Anp32; ANP32A; ANP32E; C15orf1; CPD1; HPPCn; I1PP2A; LANP; LANP-L; LANPL; MAPM; mLANP-L; PHAP1; PHAPI; PP32; W91701
UniProt ID: (Human) P39687, (Human) Q9BTT0, (Rat) P49911, (Mouse) O35381, (Mouse) P97822, (Rat) Q5XIE0
Entrez Gene ID: (Human) 8125, (Human) 81611, (Rat) 25379, (Mouse) 11737, (Mouse) 66471, (Rat) 361999
Molecular Function:
enzyme modulator
phosphatase inhibitor
phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.