Invitrogen
Manufacturer Code:PA526900
Catalog # PIPA526900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 536-565 amino acids from the C-terminal region of human PHAR4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Humdy; Phosphatase and actin regulator 4; Protein Humpty dumpty; protein phosphatase 1, regulatory subunit 124
Gene Aliases: 3110001B12Rik; AI527228; AW495572; C330013F19Rik; Kiaa4120; mKIAA4120; N28169; PHACTR4; PPP1R124; PRO2963
UniProt ID: (Human) Q8IZ21, (Mouse) Q501J7
Entrez Gene ID: (Human) 65979, (Mouse) 100169
Molecular Function: enzyme modulator phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.