Proteintech
Manufacturer Code:143271AP150UL
Catalog # 50-172-9368
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PGT Fusion Protein Ag5594 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: matrin F/G 1; OATP2A1 PGT Prostaglandin transporter SLC21A2 SLCO2A1; PGT; Prostaglandin transporter; solute carrier family 21 (prostaglandin transporter), member 2; Solute carrier family 21 member 2; Solute carrier organic anion transporter family member 2A1; solute carrier organic anion transporter family, member 2A1
Gene Aliases: 2310021C19Rik; MATR1; mPgt; OATP2A1; PGT; PHOAR2; SLC21A2; SLCO2A1
UniProt ID: (Human) Q92959, (Mouse) Q9EPT5
Entrez Gene ID: (Human) 6578, (Mouse) 24059
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.