Invitrogen
Manufacturer Code:PA543247
Catalog # PIPA543247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human PGS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDP-diacylglycerol--glycerol-3-phosphate 3-phosphatidyltransferase, mitochondrial; PGP synthase 1; Phosphatidylglycerophosphate synthase 1; Silencer-associated factor
Gene Aliases: 2610019F11Rik; 4933424M23Rik; PGS1; RGD1305052; SAF
UniProt ID: (Human) Q32NB8, (Bovine) Q2KJ28, (Mouse) Q8BHF7
Entrez Gene ID: (Human) 9489, (Bovine) 535126, (Dog) 475920, (Guinea pig) 100731635, (Mouse) 74451, (Rat) 303698
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.