Invitrogen
Manufacturer Code:PA5112945
Catalog # PIPA5112945
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human CDP-diacylglycerol--glycerol-3-phosphate 3-phosphatidyltransferase mitochondrial protein (110-400AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610019F11Rik 4933424M23Rik BOS_19570 CDP-diacylglycerol--glycerol-3-phosphate 3-phosphatidyltransferase mitochondrial PGP synthase 1 PGS1 Phosphatidylglycerophosphate synthase 1 RGD1305052 Saf Silencer-associated factor; CDP-diacylglycerol--glycerol-3-phosphate 3-phosphatidyltransferase, mitochondrial; PGP synthase 1; Phosphatidylglycerophosphate synthase 1; Silencer-associated factor
Gene Aliases: 2610019F11Rik; 4933424M23Rik; PGS1; SAF
UniProt ID: (Human) Q32NB8, (Mouse) Q8BHF7
Entrez Gene ID: (Human) 9489, (Mouse) 74451
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.