Invitrogen
Manufacturer Code:MA524166
Catalog # PIMA524166
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 439728 |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse PGRP-L Ser23-Asn530 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: HMFT0141 PGLYRPL PGRP-L PGRPL TAGL-like tagL tagL-alpha tagl-beta; N-acetylmuramoyl-L-alanine amidase; Peptidoglycan recognition protein 2; Peptidoglycan recognition protein long; PGRP-L; TagL
Gene Aliases: C730002N09Rik; Pglyrp2; Pglyrpl; PGRP-L; Pgrpl; tagL
UniProt ID: (Mouse) Q8VCS0
Entrez Gene ID: (Mouse) 57757
Molecular Function:
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.