Invitrogen
Manufacturer Code:PA579816
Catalog # PIPA579816
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human PGRMC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 25-DX; Acidic 25 kDa protein; Dap1; IZA; Membrane-associated progesterone receptor component 1; Membrane-associated progesterone receptor component 1 mPR Dap1; membrane-bound progesterone receptor; mPR; progesterone binding protein; VEMA; Ventral midline antigen
Gene Aliases: 25-Dx; 25Dx; AA415812; HPR6.6; Lewi; MPR; PGRMC; PGRMC1; PPMR; VEMA
UniProt ID: (Human) O00264, (Rat) P70580, (Mouse) O55022
Entrez Gene ID: (Human) 10857, (Rat) 291948, (Mouse) 53328
Molecular Function:
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.