Invitrogen
Manufacturer Code:PA528844
Catalog # PIPA528844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 131 and 195 of Human mPR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dap1; HPR6.6 MPR membrane-associated progesterone receptor component 1 progesterone binding protein; IZA; Membrane-associated progesterone receptor component 1; mPR; progesterone binding protein
Gene Aliases: AA415812; HPR6.6; MPR; PGRMC; PGRMC1; PPMR; Vema
UniProt ID: (Human) O00264, (Mouse) O55022
Entrez Gene ID: (Human) 10857, (Mouse) 53328
Molecular Function:
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.