Novus Biologicals
Manufacturer Code:NBP236525
Catalog # NBP236525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 145 to 195 of human Progesterone Receptor Membrane Component 1 using the numbering given in entry NP_006658.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dap1; HPR6.6PGRMC membrane-associated progesterone receptor component 1 MPR progesterone binding protein progesterone receptor membrane component 1; IZA; Membrane-associated progesterone receptor component 1; mPR; progesterone binding protein
Gene Aliases: HPR6.6; MPR; PGRMC; PGRMC1
UniProt ID: (Human) O00264
Entrez Gene ID: (Human) 10857
Molecular Function:
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.