Invitrogen
Manufacturer Code:PA528364
Catalog # PIPA528364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 209 of Human PGPEP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-oxoprolyl-peptidase; PAP-I; PGP PGP-I PGPI Pcp 5-oxoprolyl-peptidase PAP-I pyroglutamyl aminopeptidase I pyroglutamyl-peptidase 1 pyrrolidone-carboxylate peptidase; PGP-I; putative pyroglutamyl-peptidase I; Pyroglutamyl aminopeptidase I; Pyroglutamyl-peptidase 1; Pyroglutamyl-peptidase I; Pyrrolidone-carboxylate peptidase
Gene Aliases: 2810003H13Rik; Gm19458; PAP-I; Pcp; PGP; PGP-I; Pgpep1; PGPI
UniProt ID: (Mouse) Q9ESW8
Entrez Gene ID: (Mouse) 66522
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.