Invitrogen
Manufacturer Code:PA5114267
Catalog # PIPA5114267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal region of Human PGPEP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810003H13Rik 5-oxoprolyl-peptidase Gm19458 PAPI PAP-I Pcp PGI pGlu-peptidase I PGP Pgpep1 Pgpi PGP-I putative pyroglutamyl-peptidase I Pyroglutamyl aminopeptidase I Pyroglutamyl-peptidase 1 Pyroglutamyl-peptidase I Pyrrolidone-carboxylate peptidase; 5-oxoprolyl-peptidase; PAP-I; pGlu-peptidase I; PGP-I; Pyroglutamyl aminopeptidase I; Pyroglutamyl-peptidase 1; Pyroglutamyl-peptidase I; Pyrrolidone-carboxylate peptidase
Gene Aliases: PAP-I; Pcp; PGI; PGP; PGP-I; PGPEP1; PGPI
UniProt ID: (Human) Q9NXJ5
Entrez Gene ID: (Human) 54858
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.