R&D Systems
Manufacturer Code:AF6278
Catalog # AF6278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human PGPEP-1 Met1-His209 Accession # Q9NXJ5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-oxoprolyl-peptidase; EC 3.4.19.3 PAP-I PCP PGPEP1 PGPIFLJ20208 PGP-IPAP-I PGPMGC10812 Pyroglutamyl aminopeptidase I pyroglutamyl-peptidase 1 pyroglutamyl-peptidase I5-oxoprolyl-peptidase Pyrrolidone-carboxylate peptidase; PAP-I; pGlu-peptidase I; PGP-I; Pyroglutamyl aminopeptidase I; Pyroglutamyl-peptidase 1; Pyroglutamyl-peptidase I; Pyrrolidone-carboxylate peptidase
Gene Aliases: PAP-I; Pcp; PGI; PGP; PGP-I; PGPEP1; PGPI
UniProt ID: (Human) Q9NXJ5
Entrez Gene ID: (Human) 54858
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.