Invitrogen
Manufacturer Code:PA5115748
Catalog # PIPA5115748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PGP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700012G19Rik AI481330 Aspartate-based ubiquitous Mg(2+)-dependent phosphatase AUM G3PP Glycerol-3-phosphate phosphatase PGP PGPase phosphoglycolate phosphatase; Aspartate-based ubiquitous Mg(2+)-dependent phosphatase; AUM; G3PP; Glycerol-3-phosphate phosphatase; PGP; PGPase; Phosphoglycolate phosphatase
Gene Aliases: 1700012G19Rik; AI481330; AUM; G3PP; PGP; PGPase; RGD1307773
UniProt ID: (Human) A6NDG6, (Mouse) Q8CHP8, (Rat) D3ZDK7
Entrez Gene ID: (Human) 283871, (Mouse) 67078, (Rat) 287115
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.