Abnova
Manufacturer Code:H00005238M01A
Catalog # 89-105-098
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S3 |
Immunogen | PGM3 (AAH01258.11 a.a. &bsim 542 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant PGM3.
For Research Use Only
Protein Aliases: Acetylglucosamine phosphomutase; AGM1; DKFZp434B187; N-acetylglucosamine-phosphate mutase; N-acetylglucosamine-phosphate mutase 1; PAGM; PGM 3; Phosphoacetylglucosamine mutase; Phosphoglucomutase-3
Gene Aliases: AGM1; IMD23; PAGM; PGM 3; PGM3
UniProt ID: (Human) O95394
Entrez Gene ID: (Human) 5238
Molecular Function: glycosyltransferase isomerase mutase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.