OriGene
Manufacturer Code:TA504940
Catalog # 50-167-5521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5C6 |
Immunogen | Full length human recombit protein of human PGM3 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acetylglucosamine phosphomutase; AGM1 IMD23 PAGM PGM 3; N-acetylglucosamine-phosphate mutase; N-acetylglucosamine-phosphate mutase 1; PAGM; PGM 3; Phosphoacetylglucosamine mutase; Phosphoglucomutase-3
Gene Aliases: AGM1; IMD23; PAGM; PGM 3; PGM3
UniProt ID: (Human) O95394
Entrez Gene ID: (Human) 5238
Molecular Function: glycosyltransferase isomerase mutase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.