Invitrogen
Manufacturer Code:PA522353
Catalog # PIPA522353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 44 and 321 of PGM3 (Uniprot IDNo.O95394) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylglucosamine phosphomutase; AGM1 PAGM PGM 3 N-acetylglucosamine-phosphate mutase 1 acetylglucosamine phosphomutase phosphoacetylglucosamine mutase; GlcNAc-P mutase; N-acetylglucosamine-phosphate mutase; N-acetylglucosamine-phosphate mutase 1; PAGM; PGM 3; Phosphoacetylglucosamine mutase; Phosphoglucomutase-3
Gene Aliases: 2810473H05Rik; AGM1; BB187688; C77933; IMD23; PAGM; PGM 3; Pgm-3; PGM3
UniProt ID: (Human) O95394, (Mouse) Q9CYR6
Entrez Gene ID: (Human) 5238, (Mouse) 109785
Molecular Function: glycosyltransferase isomerase mutase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.