Novus Biologicals
Manufacturer Code:NBP189209
Catalog # NBP189209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:KPPQGMEIKSNERCCSFDGDADRIVYYYHDADGHFHLIDGDKIATLISSFLKELLVEIGESLNIGVVQTAYANGSSTRYLEEVMKVPVYCTKTGV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylglucosamine phosphomutase; AGM1PAGMAcetylglucosamine phosphomutase DKFZP434B187 EC 5.4.2 EC 5.4.2.3 FLJ11614 FLJ13623 N-acetylglucosamine-phosphate mutase N-acetylglucosamine-phosphate mutase 1 PGM 3 phosphoacetylglucosamine mutase phosphoglucomutase 3 Phosphoglucomutase-3; N-acetylglucosamine-phosphate mutase; N-acetylglucosamine-phosphate mutase 1; PAGM; PGM 3; Phosphoacetylglucosamine mutase; Phosphoglucomutase-3
Gene Aliases: AGM1; IMD23; PAGM; PGM 3; PGM3
UniProt ID: (Human) O95394
Entrez Gene ID: (Human) 5238
Molecular Function: glycosyltransferase isomerase mutase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.