Invitrogen
Manufacturer Code:PA544390
Catalog # PIPA544390
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PGM2L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose 1,6-bisphosphate synthase; phosphoglucomutase 2-like 1; Phosphoglucomutase 2-like 1 BM32A FLJ32029; Phosphoglucomutase-2-like 1; PMMLP
Gene Aliases: 4931406N15Rik; AI553438; BM32A; im:7140576; PGM2L1; PMMLP; zgc:198285
UniProt ID: (Human) Q6PCE3, (Mouse) Q8CAA7
Entrez Gene ID: (Bovine) 515366, (Human) 283209, (Dog) 485198, (Rabbit) 100358141, (Horse) 100052026, (Mouse) 70974, (Guinea pig) 100722597, (Rat) 685076, (Zebrafish) 449945
Molecular Function:
glycosyltransferase
isomerase
mutase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.