Novus Biologicals
Manufacturer Code:NBP19840720UL
Catalog # NBP19840720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is PGM2 - C-terminal region. Peptide sequence AIRDLTTGYDDSQPDKKAVLPTSKSSQMITFTFANGGVATMRTSGTEPKI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 5.4.2 EC 5.4.2.2 EC 5.4.2.7 FLJ10983 Glucose phosphomutase 2 PGM 2 Phosphodeoxyribomutase phosphoglucomutase 2 phosphoglucomutase-2 Phosphopentomutase; Glucose phosphomutase 2; PGM 2; Phosphodeoxyribomutase; Phosphoglucomutase-2; Phosphopentomutase
Gene Aliases: MSTP006; PGM2
UniProt ID: (Human) Q96G03
Entrez Gene ID: (Human) 55276
Molecular Function: glycosyltransferase isomerase mutase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.