Invitrogen
Manufacturer Code:PA572254
Catalog # PIPA572254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 469-501 amino acids from human PGM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose phosphomutase 1; Glucose phosphomutase 2; PGM 1; PGM 2; Phosphodeoxyribomutase; phosphoglucomutase 2; Phosphoglucomutase-1; Phosphoglucomutase-1 PGM 1 Glucose phosphomutase 1 PGM1; Phosphoglucomutase-2; Phosphopentomutase
Gene Aliases: 3230402E02Rik; CDG1T; GSD14; Pgm-1; PGM1; Pgm2
UniProt ID: (Human) P36871, (Mouse) Q7TSV4
Entrez Gene ID: (Human) 5236, (Mouse) 66681
Molecular Function:
glycosyltransferase
isomerase
mutase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.