Invitrogen
Manufacturer Code:PA544559
Catalog # PIPA544559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PGM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucose phosphomutase 1; Glucose phosphomutase 2; PGM 1; PGM 2; Phosphodeoxyribomutase; Phosphoglucomutase 1 GSD14; phosphoglucomutase 2; Phosphoglucomutase-1; Phosphoglucomutase-2; Phosphopentomutase
Gene Aliases: 3230402E02Rik; CDG1T; GSD14; PGM; Pgm-1; PGM1; Pgm2; zgc:63718
UniProt ID: (Bovine) Q08DP0, (Rabbit) P00949, (Human) P36871, (Mouse) Q7TSV4
Entrez Gene ID: (Bovine) 534402, (Dog) 479545, (Rabbit) 100009155, (Goat) 102183563, (Human) 5236, (Horse) 100069980, (Guinea pig) 100721478, (Rat) 24645, (Mouse) 66681, (Zebrafish) 394000
Molecular Function:
glycosyltransferase
isomerase
mutase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.