Novus Biologicals
Manufacturer Code:NB10056721AF594
Catalog # N106721A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 186C426 |
Immunogen | This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 95-110 of human PGRP-1 beta. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Peptidoglycan recognition protein 4; peptidoglycan recognition protein 4 peptidoglycan recognition protein I beta PGRP-I-beta; peptidoglycan recognition protein I beta; Peptidoglycan recognition protein I-beta; Peptidoglycan recognition protein intermediate beta; peptidoglycan recognition protein-I-beta; PGLYRPIbeta; PGRP-I-beta
Gene Aliases: PGLYRP4; PGLYRPIbeta; PGRP-Ibeta; PGRPIB; SBBI67
UniProt ID: (Human) Q96LB8
Entrez Gene ID: (Human) 57115
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.