Proteintech
Manufacturer Code:211341AP150UL
Catalog # 50-173-3274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PGLYRP4 Fusion Protein Ag15539 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peptidoglycan recognition protein 4; peptidoglycan recognition protein I beta; Peptidoglycan recognition protein I-beta; Peptidoglycan recognition protein intermediate beta; peptidoglycan recognition protein-I-beta; PGLYRP4 PGLYRPIbeta PGRP I beta PGRP Ibeta PGRPIB SBBI67; PGLYRPIbeta; PGRP-I-beta
Gene Aliases: PGLYRP4; PGLYRPIbeta; PGRP-Ibeta; PGRPIB; SBBI67
UniProt ID: (Human) Q96LB8
Entrez Gene ID: (Human) 57115
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.