Novus Biologicals
Manufacturer Code:NBP180919
Catalog # NBP180919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:PAALSAAEGLISYAIQKGHLSPRYIQPLLLKEETCLDPQHPVMPRKVCPNIIKRSAWEARETHCPK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peptidoglycan recognition protein 3; peptidoglycan recognition protein 3 peptidoglycan recognition protein I alpha Peptidoglycan recognition protein I-alpha Peptidoglycan recognition protein intermediate alpha peptidoglycan recognition protein-I-alpha PGLYRPIalphaMGC149197 PGRP-Ialpha PGRPIAPGRP-I-alpha; peptidoglycan recognition protein I alpha; Peptidoglycan recognition protein I-alpha; Peptidoglycan recognition protein intermediate alpha; PGLYRPIalpha
Gene Aliases: PGLYRP3; PGLYRPIalpha; PGRP-Ialpha; PGRPIA
UniProt ID: (Human) Q96LB9
Entrez Gene ID: (Human) 114771
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.