R&D Systems
Manufacturer Code:IC4704V100UG
Catalog # IC4704V100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 439728 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse PGLYRP2/PGRP-L Ser23-Asn530 Accession # Q8VCS0 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: EC 3.5.1.28 peptidoglycan recognition protein 2tagl-beta peptidoglycan recognition protein L PGLYRPL PGLYRPLHMFT0141 PGRP2 PGRPL PGRP-L PGRP-LN-acetylmuramoyl-L-alanine amidase PGRPLPeptidoglycan recognition protein long tagL tagL-alpha TAGL-like; N-acetylmuramoyl-L-alanine amidase; Peptidoglycan recognition protein 2; peptidoglycan recognition protein L; Peptidoglycan recognition protein long; PGRP-L
Gene Aliases: HMFT0141; PGLYRP2; PGLYRPL; PGRP-L; PGRPL; tagL; tagL-alpha; tagl-beta; TAGL-like; UNQ3103/PRO10102
UniProt ID: (Human) Q96PD5
Entrez Gene ID: (Human) 114770
Molecular Function:
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.