Novus Biologicals
Manufacturer Code:NB10056719PECY55
Catalog # N1056719PC5
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 188C424 |
Immunogen | This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 165-180 (YVLKGHRDVQRTLSPG) of human PGRP-S (NP_005082). |
Conjugate | PE-Cyanine5.5 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: Peptidoglycan recognition protein 1; peptidoglycan recognition protein 1 Peptidoglycan recognition protein short PGLYRP PGRPpeptidoglycan recognition protein PGRP-SMGC126894 PGRPSMGC126896 TAG7 TNF superfamily member 3 (LTB)-like (peptidoglycan recognition protein) TNFSF3L; Peptidoglycan recognition protein short; PGRP-S; TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein)
Gene Aliases: PGLYRP; PGLYRP1; PGRP; PGRP-S; PGRPS; SBBI68; TAG7; TNFSF3L; UNQ639/PRO1269
UniProt ID: (Human) O75594
Entrez Gene ID: (Human) 8993
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.