R&D Systems
Manufacturer Code:BAF2590
Catalog # BAF2590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human PGLYRP1/PGRP-S Gln22-Pro196 Accession # O75594 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Peptidoglycan recognition protein 1; peptidoglycan recognition protein 1 Peptidoglycan recognition protein short PGLYRP PGRPpeptidoglycan recognition protein PGRPS PGRP-S PGRP-SMGC126894 PGRPSMGC126896 TAG7 TNF superfamily member 3 (LTB)-like (peptidoglycan recognition protein) TNFSF3L; Peptidoglycan recognition protein short; PGRP-S; TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein)
Gene Aliases: PGLYRP; PGLYRP1; PGRP; PGRP-S; PGRPS; SBBI68; TAG7; TNFSF3L; UNQ639/PRO1269
UniProt ID: (Human) O75594
Entrez Gene ID: (Human) 8993
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.