Invitrogen
Manufacturer Code:PA544901
Catalog # PIPA544901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PGLS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphogluconolactonase; 6-phosphogluconolactonase {ECO:0000250|UniProtKB:O95336}; 6PGL; 6PGL {ECO:0000250|UniProtKB:O95336}; epididymis secretory protein Li 304; pgls {ECO:0000250|UniProtKB:O95336}
Gene Aliases: 1110030K05Rik; 6PGL; AI447866; HEL-S-304; PGLS; Plgs
UniProt ID: (Bovine) Q2TBQ8, (Human) O95336, (Mouse) Q9CQ60
Entrez Gene ID: (Horse) 100070210, (Bovine) 616120, (Pig) 100517636, (Human) 25796, (Dog) 610090, (Mouse) 66171, (Rat) 290636, (Guinea pig) 100734971
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.