Novus Biologicals
Manufacturer Code:NBP15638720UL
Catalog # NBP15638720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PGLS(6-phosphogluconolactonase) The peptide sequence was selected from the middle region of PGLS. Peptide sequence AAVLKRILEDQEENPLPAALVQPHTGKLCWFLDEAAARLLTVPFEKHSTL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphogluconolactonase; 6PGL; 6PGLEC 3.1.1.31 6-phosphogluconolactonase; epididymis secretory protein Li 304
Gene Aliases: 6PGL; HEL-S-304; PGLS
UniProt ID: (Human) O95336
Entrez Gene ID: (Human) 25796
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.