Invitrogen
Manufacturer Code:PA556529
Catalog # PIPA556529
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PGGT1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTC-428G20.3; Geranylgeranyl transferase type I subunit beta; Geranylgeranyl transferase type-1 subunit beta; geranylgeranyltransferase type 1 beta; geranylgeranyltransferase type I (GGTase-I); geranylgeranyltransferase type I beta subunit; GGTase-I-beta; protein geranylgeranyltransferase type I, beta subunit; Type I protein geranyl-geranyltransferase subunit beta
Gene Aliases: 2010207C17Rik; 2610100E13; AI451237; AI551093; BGG1; BGGI; GGT1; GGTI; PGGT1B
UniProt ID: (Human) P53609, (Mouse) Q8BUY9, (Rat) P53610
Entrez Gene ID: (Human) 5229, (Mouse) 225467, (Rat) 81746
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.