Novus Biologicals
Manufacturer Code:NBP2854670.1ML
Catalog # NB033396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Rat PGGT1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BGGI EC 2.5.1 EC 2.5.1.59 Geranylgeranyl transferase type I subunit beta geranylgeranyl transferase type-1 subunit beta geranylgeranyltransferase type I beta subunit GGTase-I-beta GGTI protein geranylgeranyltransferase type I beta subunit Type I protein geranyl-geranyltransferase subunit beta; CTC-428G20.3; Geranylgeranyl transferase type I subunit beta; Geranylgeranyl transferase type-1 subunit beta; geranylgeranyltransferase type I beta subunit; GGTase-I-beta; protein geranylgeranyltransferase type I, beta subunit; Type I protein geranyl-geranyltransferase subunit beta
Gene Aliases: BGGI; GGTI; PGGT1B
UniProt ID: (Human) P53609
Entrez Gene ID: (Human) 5229
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.