Novus Biologicals
Manufacturer Code:NB11059911AF532
Catalog # N1109911A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The epitope recognized by this antibody maps to region within residue 120-190 of human Prostaglandin D Synthase/Lipocalin. [Swiss-ProtNo. P41222] |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: Beta-trace protein; Beta-trace protein cerebrin-28 EC 5.3.99.2 Glutathione-independent PGD synthase glutathione-independent PGD synthetase lipocalin-type prostaglandin D synthase Lipocalin-type prostaglandin-D synthase L-PGDS PDS PGD2 PGD2 synthase PGDS2 PGDSLPGDS prostaglandin D synthase prostaglandin D2 synthase (21kD brain) prostaglandin D2 synthase 21kDa (brain) Prostaglandin-D2 synthase prostaglandin-H2 D-isomerase; Cerebrin-28; Glutathione-independent PGD synthase; glutathione-independent PGD synthetase; L-PGDS; lipocalin-type prostaglandin D synthase; Lipocalin-type prostaglandin-D synthase; PGD2 synthase; prostaglandin D synthase; prostaglandin D2 synthase 21kDa (brain); Prostaglandin-D2 synthase; Prostaglandin-H2 D-isomerase; testis tissue sperm-binding protein Li 63n
Gene Aliases: L-PGDS; LPGDS; PDS; PGD2; PGDS; PGDS2; PTGDS
UniProt ID: (Human) P41222
Entrez Gene ID: (Human) 5730
Molecular Function: isomerase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.