Novus Biologicals
Manufacturer Code:NBP273361F
Catalog # NBP273361F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A5 |
Immunogen | Full length recombinant protein of human PGD (NP_002622) produced in HEK293T cells. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 6-phosphogluconate dehydrogenase, decarboxylating; 6PGD EC 1.1.1.446-phosphogluconate dehydrogenase decarboxylating PGDH phosphogluconate dehydrogenase
Gene Aliases: 6PGD; PGD; PGDH
UniProt ID: (Human) P52209
Entrez Gene ID: (Human) 5226
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.