Invitrogen
Manufacturer Code:PA588327
Catalog # PIPA588327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CPQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aminopeptidase; blood plasma glutamate carboxypeptidase; Carboxypeptidase Q; Hematopoietic lineage switch 2; Hematopoietic lineage switch 2 related protein; Hls2-rp; LAL-1; liver annexin like-1; Liver annexin-like protein 1; Lysosomal dipeptidase; Plasma glutamate carboxypeptidase; Ser-Met dipeptidase
Gene Aliases: 1190003P12Rik; 2610034C17Rik; CPQ; Hls2; LAL; Lal-1; LCH1; LDP; PGCP
UniProt ID: (Human) Q9Y646, (Rat) Q6IRK9, (Mouse) Q9WVJ3
Entrez Gene ID: (Human) 10404, (Rat) 58952, (Mouse) 54381
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.