Invitrogen
Manufacturer Code:PA547602
Catalog # PIPA547602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse PGCP Lys19-Ser470 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: aminopeptidase; Carboxypeptidase Q; Hematopoietic lineage switch 2; Plasma glutamate carboxypeptidase
Gene Aliases: 1190003P12Rik; 2610034C17Rik; Cpq; Hls2; Lal-1; Pgcp
UniProt ID: (Mouse) Q9WVJ3
Entrez Gene ID: (Mouse) 54381
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.