Invitrogen
Manufacturer Code:PA5113866
Catalog # PIPA5113866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal region of human PGCP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1190003P12Rik 2610034C17Rik aminopeptidase blood plasma glutamate carboxypeptidase Carboxypeptidase Q CPQ Hematopoietic lineage switch 2 Hematopoietic lineage switch 2 related protein Hls2 Hls2-rp LAL LAL-1 LCH1 LDP liver annexin like-1 Liver annexin-like protein 1 Lysosomal dipeptidase Pgcp Plasma glutamate carboxypeptidase Ser-Met dipeptidase; aminopeptidase; blood plasma glutamate carboxypeptidase; Carboxypeptidase Q; Lysosomal dipeptidase; Plasma glutamate carboxypeptidase; Ser-Met dipeptidase
Gene Aliases: CPQ; LCH1; LDP; PGCP
UniProt ID: (Human) Q9Y646
Entrez Gene ID: (Human) 10404
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.