R&D Systems
Manufacturer Code:AF4258SP
Catalog # AF4258SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse PGCP Lys19-Ser470 Accession # Q9WVJ3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: aminopeptidase; aminopeptidase CPQ EC 3.4.17 EC 3.4.17.- Hls2 Lal-1 plasma glutamate carboxypeptidase; blood plasma glutamate carboxypeptidase; Carboxypeptidase Q; Lysosomal dipeptidase; Plasma glutamate carboxypeptidase; Ser-Met dipeptidase
Gene Aliases: CPQ; LCH1; LDP; PGCP
UniProt ID: (Human) Q9Y646
Entrez Gene ID: (Human) 10404
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.