Novus Biologicals
Manufacturer Code:NBP237562
Catalog # NBP237562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G8 |
Immunogen | This PGC1 alpha Antibody (1G8) was developed against a purified recombinant fragment of human PGC-1alpha expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: L-PGC-1alpha; LEM6 Ligand effect modulator 6 ligand effect modulator-6 L-PGC-1alpha peroxisome proliferative activated receptor gamma coactivator 1 alpha peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT peroxisome proliferator-activated receptor gamma coactivator 1-alpha peroxisome proliferator-activated receptor gamma coactivator 1 alpha PGC1 PGC-1(alpha) PGC1A PGC-1-alpha PGC1APGC-1(alpha) PGC1peroxisome proliferative activated receptor gamma coactivator 1 PGC-1v PPAR gamma coactivator variant form PPAR gamma coactivator-1 PPARgamma coactivator 1alpha PPAR-gamma coactivator 1-alpha PPARGC1 PPARGC-1-alpha; Ligand effect modulator 6; ligand effect modulator-6; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT; Peroxisome proliferator-activated receptor gamma coactivator 1-alpha; peroxisome proliferator-activated receptor gamma, coactivator 1 alpha; PGC-1-alpha; PPAR gamma coactivator variant form; PPARgamma coactivator 1alpha; PPARGC-1-alpha
Gene Aliases: LEM6; PGC-1(alpha); PGC-1alpha; PGC-1v; PGC1; PGC1A; PPARGC1; PPARGC1A
UniProt ID: (Human) Q3LIG1
Entrez Gene ID: (Human) 10891
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.