Invitrogen
Manufacturer Code:PA538021
Catalog # PIPA538021
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 19 amino acid peptide near the carboxy terminus of human PPARGC1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: L-PGC-1alpha; Ligand effect modulator 6; ligand effect modulator-6; peroxisome proliferative activated receptor gamma coactivator 1; peroxisome proliferative activated receptor, gamma, coactivator 1 alpha; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT; Peroxisome proliferator-activated receptor gamma coactivator 1-alpha; peroxisome proliferator-activated receptor gamma coactivator-1 alpha; peroxisome proliferator-activated receptor gamma, coactivator 1 alpha; PGC-1-alpha; PGC-1alpha; Pgc-1alphaa; PPAR gamma coactivator variant form; PPAR Gamma Coactivator-1; PPAR-gamma coactivator 1-alpha; PPARgamma coactivator 1alpha; PPARGC-1-alpha; PPARGC1A LEM6 PGC1 PGC1A PGC-1v PPARGC1 LEM6 Peroxisome proliferator-activated receptor gamma coactivator 1-alpha Ligand effect modulator 6 PGC-1-alpha
Gene Aliases: A830037N07Rik; ENSMUSG00000079510; Gm11133; LEM6; PGC-1; PGC-1(alpha); PGC-1alpha; PGC-1v; PGC1; PGC1A; Pgco1; PPARGC1; PPARGC1A
UniProt ID: (Human) Q3LIG1, (Mouse) O70343, (Rat) Q9QYK2
Entrez Gene ID: (Human) 10891, (Mouse) 19017, (Rat) 83516
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.