Novus Biologicals
Manufacturer Code:NBP104676AF700
Catalog # NBP104676Z
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This PGC1 alpha Antibody was developed against a synthetic peptide made to an internal portion of the human PGC-1 alpha protein (within residues 400-550). [Swiss-Prot# Q9UBK2]. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: L-PGC-1alpha; LEM6 Ligand effect modulator 6 ligand effect modulator-6 L-PGC-1alpha peroxisome proliferative activated receptor gamma coactivator 1 alpha peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT peroxisome proliferator-activated receptor gamma coactivator 1-alpha peroxisome proliferator-activated receptor gamma coactivator 1 alpha PGC1 PGC-1(alpha) PGC1A PGC-1-alpha PGC1APGC-1(alpha) PGC1peroxisome proliferative activated receptor gamma coactivator 1 PGC-1v PPAR gamma coactivator variant form PPAR gamma coactivator-1 PPARgamma coactivator 1alpha PPAR-gamma coactivator 1-alpha PPARGC1 PPARGC-1-alpha; Ligand effect modulator 6; ligand effect modulator-6; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT; Peroxisome proliferator-activated receptor gamma coactivator 1-alpha; peroxisome proliferator-activated receptor gamma, coactivator 1 alpha; PGC-1-alpha; PPAR gamma coactivator variant form; PPARgamma coactivator 1alpha; PPARGC-1-alpha
Gene Aliases: LEM6; PGC-1(alpha); PGC-1alpha; PGC-1v; PGC1; PGC1A; PPARGC1; PPARGC1A
UniProt ID: (Human) Q3LIG1
Entrez Gene ID: (Human) 10891
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.