Novus Biologicals
Manufacturer Code:NBP169334
Catalog # NBP169334
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PERLD1(per1-like domain containing 1) The peptide sequence was selected from the N terminal of PERLD1. Peptide sequence AGLAARLVLLAGAAALASGSQGDREPVYRDCVLQCEEQNCSGGALNHFRS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGLA546 CAB2COS16 homolog Gene coamplified with ERBB2 protein MGC9753hCOS16 PER1 per1-like domain containing 1 PERLD1PER1-like domain-containing protein 1 post-GPI attachment to proteins 3 post-GPI attachment to proteins factor 3 PP1498; COS16 homolog; Gene coamplified with ERBB2 protein; hCOS16; per1-like domain containing 1; PER1-like domain-containing protein 1; Post-GPI attachment to proteins factor 3
Gene Aliases: AGLA546; CAB2; hCOS16; PERLD1; PGAP3; PP1498; UNQ546/PRO1100
UniProt ID: (Human) Q8NBJ8
Entrez Gene ID: (Human) 93210
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.