Invitrogen
Manufacturer Code:PA572340
Catalog # PIPA572340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 90-122 amino acids from human PGAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GPI deacylase; GPI inositol-deacylase; GPI inositol-deacylase 3.1.-.- Post-GPI attachment to proteins factor 1 hPGAP1 PGAP1; hPGAP1; Post GPI Attachment to Proteins 1; Post-GPI attachment to proteins factor 1
Gene Aliases: 5033403E17Rik; 9030223K07Rik; A530084K22; Bst1; D230012E17Rik; ISPD3024; MRT42; oto; PGAP1; SPG67; UNQ3024/PRO9822
UniProt ID: (Human) Q75T13, (Mouse) Q3UUQ7, (Rat) Q765A7
Entrez Gene ID: (Human) 80055, (Mouse) 241062, (Rat) 316400
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.