Abnova
Manufacturer Code:H00080055A01
Catalog # 89-107-355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 168 a.a. &bsim 266 a.a) partial recombinant protein with GST tag.,PGAP1 (NP_079265 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PGAP1.
For Research Use Only
Protein Aliases: Bst1; FLJ42774; GPI inositol-deacylase; hPGAP1; ISPD3024; Post-GPI attachment to proteins factor 1
Gene Aliases: Bst1; ISPD3024; MRT42; PGAP1; SPG67; UNQ3024/PRO9822
UniProt ID: (Human) Q75T13
Entrez Gene ID: (Human) 80055
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.