Invitrogen
Manufacturer Code:MA527360
Catalog # PIMA527360
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A1 |
Immunogen | Full length human recombit protein of PGAM4 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: PGAM1 PGAM2 PGAM4 PGAM1 PGAM2 phosphoglycerate mutase 1 PGAMA RP11-452K12.8 OTTHUMP00000059414 Phosphoglycerate mutase A phosphoglycerate mutase 2 MGC88743 PGAMM Phosphoglycerate mutase muscle PGAM3 phosphoglycerate mutase family member 4 PGAM-B dJ1000K24.1 phosphoglycerate mutase family 3 phosphoglycerate mutase family 4 phosphoglycerate mutase processed protein bisphosphoglycerate mutase 1 bisphosphoglycerate mutase 2 bisphosphoglycerate mutase 4; phosphoglycerate mutase family 3; phosphoglycerate mutase processed protein; Probable phosphoglycerate mutase 4
Gene Aliases: dJ1000K24.1; PGAM-B; PGAM1; PGAM3; PGAM4
UniProt ID: (Human) Q8N0Y7
Entrez Gene ID: (Human) 441531
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.