Invitrogen
Manufacturer Code:PA524006
Catalog # PIPA524006
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 12-41 amino acids from the N-terminal region of human PGAM2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BPG-dependent PGAM 2; GSD10 PGAM-M PGAMM BPG-dependent PGAM 2 muscle-specific phosphoglycerate mutase phosphoglycerate mutase 2 phosphoglycerate mutase isozyme M; Muscle-specific phosphoglycerate mutase; PGAM-M; Phosphoglycerate mutase 2; phosphoglycerate mutase 2 (muscle); Phosphoglycerate mutase isozyme M; phosphoglycerate mutase muscle-specific subunit
Gene Aliases: GSD10; PGAM-M; PGAM2; PGAMM
UniProt ID: (Human) P15259, (Mouse) O70250
Entrez Gene ID: (Human) 5224, (Mouse) 56012
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.