Invitrogen
Manufacturer Code:PA517935
Catalog # PIPA517935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KAMEAVAAQGKAKK) corresponding to the C-terminus amino acids of PGAM1 / PGAM2 / PGAM4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BPG-dependent PGAM 1; BPG-dependent PGAM 2; epididymis secretory protein Li 35; Muscle-specific phosphoglycerate mutase; PGAM-B; PGAM-M; PGAM1 PGAM2 PGAM4 PGAM1 PGAM2 phosphoglycerate mutase 1 PGAMA RP11-452K12.8 OTTHUMP00000059414 Phosphoglycerate mutase A phosphoglycerate mutase 2 MGC88743 PGAMM Phosphoglycerate mutase muscle PGAM3 phosphoglycerate mutase family member 4 PGAM-B dJ1000K24.1 phosphoglycerate mutase family 3 phosphoglycerate mutase family 4 phosphoglycerate mutase processed protein bisphosphoglycerate mutase 1 bisphosphoglycerate mutase 2 bisphosphoglycerate mutase 4; Phosphoglycerate mutase 1; phosphoglycerate mutase 1 (brain); Phosphoglycerate mutase 2; phosphoglycerate mutase 2 (muscle); phosphoglycerate mutase A, nonmuscle form; phosphoglycerate mutase family 3; Phosphoglycerate mutase isozyme B; Phosphoglycerate mutase isozyme M; phosphoglycerate mutase muscle-specific subunit; phosphoglycerate mutase processed protein; Probable phosphoglycerate mutase 4
Gene Aliases: CDABP0006; D14Mgh1; dJ1000K24.1; GSD10; HEL-S-35; PGAM-B; PGAM-M; PGAM1; PGAM2; PGAM3; PGAM4; PGAMA; PGAMM; Pgmut
UniProt ID: (Human) P18669, (Human) P15259, (Human) Q8N0Y7, (Mouse) O70250, (Rat) P16290
Entrez Gene ID: (Human) 5223, (Human) 5224, (Human) 441531, (Mouse) 56012, (Rat) 24959
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.