Invitrogen
Manufacturer Code:PA5115744
Catalog # PIPA5115744
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PGAM2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310050F24Rik BPG-dependent PGAM 1 BPG-dependent PGAM 2 CDABP0006 D14Mgh1 dJ1000K24.1 epididymis secretory protein Li 35 GSD10 HEL-S-35 MGC88743 Muscle-specific phosphoglycerate mutase OTTHUMP00000059414 PGAM1 Pgam-1 Pgam2 PGAM3 PGAM4 PGAMA PGAM-B PGAMM PGAM-M Pgmut Phosphoglycerate mutase 1 phosphoglycerate mutase 1 (brain) phosphoglycerate mutase 2 phosphoglycerate mutase 2 (muscle) phosphoglycerate mutase 4 phosphoglycerate mutase A nonmuscle form phosphoglycerate mutase family 3 phosphoglycerate mutase family member 4 phosphoglycerate mutase isozyme B Phosphoglycerate mutase isozyme M phosphoglycerate mutase muscle-specific subunit phosphoglycerate mutase processed protein probable phosphoglycerate mutase 4 RP11-452K12.8; BPG-dependent PGAM 1; BPG-dependent PGAM 2; epididymis secretory protein Li 35; Muscle-specific phosphoglycerate mutase; PGAM-B; PGAM-M; Phosphoglycerate mutase 1; phosphoglycerate mutase 1 (brain); Phosphoglycerate mutase 2; phosphoglycerate mutase 2 (muscle); phosphoglycerate mutase A, nonmuscle form; phosphoglycerate mutase family 3; Phosphoglycerate mutase isozyme B; Phosphoglycerate mutase isozyme M; phosphoglycerate mutase muscle-specific subunit; phosphoglycerate mutase processed protein; Probable phosphoglycerate mutase 4
Gene Aliases: 2310050F24Rik; CDABP0006; D14Mgh1; dJ1000K24.1; GSD10; HEL-S-35; Pgam-1; PGAM-B; PGAM-M; PGAM1; PGAM2; PGAM3; PGAM4; PGAMA; PGAMM; Pgmut
UniProt ID: (Human) P18669, (Human) P15259, (Human) Q8N0Y7, (Rat) P25113, (Mouse) Q9DBJ1, (Mouse) O70250, (Rat) P16290
Entrez Gene ID: (Human) 5223, (Human) 5224, (Human) 441531, (Rat) 24642, (Mouse) 18648, (Mouse) 56012, (Rat) 24959
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.