Invitrogen
Manufacturer Code:PA583617
Catalog # PIPA583617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PGAM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BPG-dependent PGAM 1; epididymis secretory protein Li 35; PGAM-B; PGAM-B PGAMA; Phosphoglycerate mutase 1; phosphoglycerate mutase 1 (brain); phosphoglycerate mutase A, nonmuscle form; Phosphoglycerate mutase isozyme B
Gene Aliases: CDABP0006; HEL-S-35; PGAM-B; PGAM1; PGAMA
UniProt ID: (Human) P18669
Entrez Gene ID: (Human) 5223
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.